Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TROY/TNFRSF19 Antibody (104), HRP , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290450H
Description
TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Mouse samples. It is validated for ELISA, ELISASpecifications
TROY/TNFRSF19 | |
Monoclonal | |
HRP | |
TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TROY/TNFRSF19 (NP_001157627.1; Met1-Leu170). | |
0.1 mL | |
Apoptosis | |
55504 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
104 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction