Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TROY/TNFRSF19 Antibody (933214), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB1548PE
Description
TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry.Specifications
TROY/TNFRSF19 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
TNFRSF19 | |
Partial recombinant human TROY/TNFRSF protein (19 aa30-170) | |
100 μL | |
Apoptosis | |
55504 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
933214 | |
Flow Cytometry | |
TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction