Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TROY/TNFRSF19 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen TROY/TNFRSF19
Concentration 0.5 mg/mL
Dilution Western Blot 0.5-2 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB12012126T
SDP
View Documents
Novus Biologicals
NB12012126SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB12012126
SDP
View Documents
Novus Biologicals
NB12012126
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

TROY/TNFRSF19 Polyclonal specifically detects TROY/TNFRSF19 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TROY/TNFRSF19
Western Blot 0.5-2 ug/ml
Polyclonal
Purified
RUO
Human, Mouse
Q9NS68
55504
This rabbit polyclonal antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 29-44, 204-219, and 409-421 of human TROY (Genbank Accession No. AB040434).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Apoptosis
PBS containing 0.05% BSA with 0.05% Sodium Azide
TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
TNFRSF19
IgG
Protein G purified
Cross-reacts with Human and Mouse. Not yet tested in other species.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.