Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPA1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5097207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11040763T 0.025 mL
NB11040763 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11040763T Supplier Novus Biologicals Supplier No. NB11040763SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 41 publication

TRPA1 Polyclonal specifically detects TRPA1 in Human, Mouse, Rat, Guinea Pig, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TRPA1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 28990934), ELISA reported in scientific literature (PMID 29803505), Immunohistochemistry 0.5-1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:100-1:250, Immunohistochemistry-Frozen reported in scientific literature
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O75762
Gene Alias ANKTM1ankyrin-like with transmembrane domains 1, Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, transient receptor potential cation channel subfamily A member 1, transient receptor potential cation channel, subfamily A, member 1
Gene Symbols TRPA1
Host Species Rabbit
Immunogen A synthetic peptide made to a region within the N-terminus (residues 1-100) of the human TRPA1 protein. [Swiss-Prot# O75762]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems
Primary or Secondary Primary
Gene ID (Entrez) 8989
Test Specificity Additional modified form of TRPA1 can also be detected.
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.