Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPA1 Antibody, Novus Biologicals™
SDP

Catalog No. NB017954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017954 0.025 mL
NB10098836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017954 Supplier Novus Biologicals Supplier No. NB100988360.025ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPA1 Polyclonal specifically detects TRPA1 in Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TRPA1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Whole antisera with No Preservative
Gene Alias ANKTM1ankyrin-like with transmembrane domains 1, Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, transient receptor potential cation channel subfamily A member 1, transient receptor potential cation channel, subfamily A, member 1
Gene Symbols TRPA1
Host Species Rabbit
Immunogen A synthetic peptide from aa region 1-50 of drosophila TRPA1 conjugated to an immunogenic carrier protein was used as the antigen.
Molecular Weight of Antigen 145 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8989
Test Specificity TRPA1.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration.
Target Species Drosophila
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.