Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$206.50 - $501.50
Specifications
Antigen | TRPA1 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017898
![]() |
Novus Biologicals
NB100913190.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NB10091319
![]() |
Novus Biologicals
NB10091319 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
TRPA1 Polyclonal specifically detects TRPA1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TRPA1 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
ANKTM1ankyrin-like with transmembrane domains 1, Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, transient receptor potential cation channel subfamily A member 1, transient receptor potential cation channel, subfamily A, member 1 | |
TRPA1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
Q6RI86 | |
8989 | |
IgG | |
Unpurified | |
128 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title