Learn More
TRPA1 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111755
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)NSTGIIN ETSDHSE Homology: Bovine amino acid residues identical; canis - 13/14 amino acid residues identical; rat - 12/14 amino acid residues identical; mouse - 9/14 amino acid residues identical.
TRPA1 is a TRP-related channel that responds to cold temperatures and pungent compounds and plays a role in both nociceptor and hair cell transduction. TRPA1 is a transformation -associated gene product in lung epithelia, whereas its protein distribution is primarily restricted to sensory neurons. Blocking TRPA1 may be a therapeutic target for treating cold hyperalgesia caused by inflammation and nerve damage. It is now known that TRPA1 protein is also widely expressed outside of the CNS and is dys-regulated during oncogenic transformation. The structure of TRPA1 is highly related to both the protein ankyrin and transmembrane proteins. The specific function of TRPA1 has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control.
Specifications
TRPA1 (extracellular) | |
Polyclonal | |
Unconjugated | |
Trpa1 | |
ANKTM1; Ankyrin-like with transmembrane domains protein 1; FEPS; nompC; Transformation-sensitive protein p120; Transient receptor potential cation channel subfamily A member 1; Trpa1; TRPA-1; TRPA1b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
277328, 312896, 8989 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O75762, Q6RI86, Q8BLA8 | |
Trpa1 | |
Peptide corresponding to amino acidresidues 747-760 of human TRPA1 (1stextracellular loop) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.