Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533089
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533089 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533089 Supplier Invitrogen™ Supplier No. PA533089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey (100%); Marmoset (94%); Dog, Bovine, Elephant (89%); Rat, Bat, Panda, Horse, Rabbit (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

TRPA1 is a TRP-related channel that responds to cold temperatures and pungent compounds and plays a role in both nociceptor and hair cell transduction. TRPA1 is a transformation -associated gene product in lung epithelia, whereas its protein distribution is primarily restricted to sensory neurons. Blocking TRPA1 may be a therapeutic target for treating cold hyperalgesia caused by inflammation and nerve damage. It is now known that TRPA1 protein is also widely expressed outside of the CNS and is dys-regulated during oncogenic transformation. The structure of TRPA1 is highly related to both the protein ankyrin and transmembrane proteins. The specific function of TRPA1 has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPA1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Trpa1
Gene Accession No. O75762
Gene Alias Anktm1; ankyrin-like with transmembrane domains 1; ankyrin-like with transmembrane domains protein 1; FEPS; ion channel; drTRPA1; nompC; transformation-sensitive protein p120; transient receptor potential A1; transient receptor potential ankyrin 1a; transient receptor potential cation channel subfamily A member 1; transient receptor potential cation channel subfamily A member 1a; transient receptor potential cation channel, subfamily A, member 1a; transient receptor potential cation channel subfamily A member 1-like; transient receptor potential cation channel, subfamily A, member 1; transient receptor potential cation channel, subfamily A, member 1 L homeolog; transient receptor potential cation channel, subfamily A, member 1a; TRPA1; TRPA-1; trpa1.L; trpa1a; TRPA1b; XELAEV_18003616mg
Gene Symbols Trpa1
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human TRPA1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8989
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.