Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109320
Description
Positive Control: L929, MG-63, NIH-3T3, rat brain tissue, human skin tissue, human esophagus tissue, mouse colon tissue.
TRPA1 is a TRP-related channel that responds to cold temperatures and pungent compounds and plays a role in both nociceptor and hair cell transduction. TRPA1 is a transformation -associated gene product in lung epithelia, whereas its protein distribution is primarily restricted to sensory neurons. Blocking TRPA1 may be a therapeutic target for treating cold hyperalgesia caused by inflammation and nerve damage. It is now known that TRPA1 protein is also widely expressed outside of the CNS and is dys-regulated during oncogenic transformation. The structure of TRPA1 is highly related to both the protein ankyrin and transmembrane proteins. The specific function of TRPA1 has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control.
Specifications
| TRPA1 | |
| Polyclonal | |
| Unconjugated | |
| Trpa1 | |
| Anktm1; ankyrin-like with transmembrane domains 1; ankyrin-like with transmembrane domains protein 1; FEPS; ion channel; drTRPA1; nompC; transformation-sensitive protein p120; transient receptor potential A1; transient receptor potential ankyrin 1a; transient receptor potential cation channel subfamily A member 1; transient receptor potential cation channel subfamily A member 1a; transient receptor potential cation channel, subfamily A, member 1a; transient receptor potential cation channel subfamily A member 1-like; transient receptor potential cation channel, subfamily A, member 1; transient receptor potential cation channel, subfamily A, member 1 L homeolog; transient receptor potential cation channel, subfamily A, member 1a; TRPA1; TRPA-1; trpa1.L; trpa1a; TRPA1b; XELAEV_18003616mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8989 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| O75762 | |
| Trpa1 | |
| Recombinant protein within Human TRPA1 aa 505-755. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction