Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$501.50
Specifications
Antigen | TRPA1 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TRPA1 Polyclonal specifically detects TRPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TRPA1 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems | |
PBS buffer, 2% sucrose | |
8989 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ANKTM1ankyrin-like with transmembrane domains 1, Ankyrin-like with transmembrane domains protein 1, Transformation-sensitive protein p120, transient receptor potential cation channel subfamily A member 1, transient receptor potential cation channel, subfamily A, member 1 | |
The immunogen is a synthetic peptide directed towards the middle region of human TRPA1 (NP_015628). Peptide sequence KCTDRLDEDGNTALHFAAREGHAKAVALLLSHNADIVLNKQQASFLHLAL | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title