Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC1 Antibody (SR1100), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32202950UL
Description
TRPC1 Monoclonal antibody specifically detects TRPC1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TRPC1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1100 | |
Western Blot 1:500-1:2000 | |
HTRP-1, MGC133334, MGC133335, transient receptor potential canonical 1, transient receptor potential cation channel, subfamily C, member 1, transient receptor potential channel 1, Transient receptor protein 1, TRP-1, TRP1short transient receptor potential channel 1, TrpC1 | |
A synthesized peptide derived from human TRPC1 (Uniprot #: P48995) | |
50 μg | |
Cardiovascular Biology, Neuroscience, Signal Transduction | |
7220 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction