Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110525 Supplier Invitrogen™ Supplier No. PA5110525
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLNLYSLVYN EDKKNTMGPA LERIDYLLIL WIIGMIWSDI KRLWYEGLED FLEESRNQLS FVMNSLYLAT FALKVVAHNK FHDFADRKDW DAFHPTLVAE G.

Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Trpc1
Gene Accession No. P48995, Q61056
Gene Alias capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; Mtrp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; transient receptor protein 1; trp; TRP1; TRP-1; trpc1; trpc1.L; trp-related protein 1; Trrp1; XELAEV_18027727mg; xtrpc1
Gene Symbols Trpc1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400-500 of human TRPC1 (NP_001238774.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22063, 7220
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.