Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100358
Description
Antibody detects endogenous levels of total TRPC1.
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.
Specifications
TRPC1 | |
Polyclonal | |
Unconjugated | |
Trpc1 | |
capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; Mtrp1; Short transient receptor potential channel 1; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; transient receptor protein 1; trp; TRP1; TRP-1; trpc1; trpc1.L; trp-related protein 1; Trrp1; XELAEV_18027727mg; xtrpc1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
22063, 7220, 89821 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P48995, Q61056, Q9QX01 | |
Trpc1 | |
A synthesized peptide derived from human TRPC1(Accession P48995), corresponding to amino acid residues I695-S745. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction