Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC1 Rabbit anti-Human, Mouse, Rat, Clone: 1V3Q2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | TRPC1 |
---|---|
Clone | 1V3Q2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TRPC1 Monoclonal antibody specifically detects TRPC1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TRPC1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7220 | |
IgG | |
Affinity purified |
1V3Q2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
HTRP-1, MGC133334, MGC133335, transient receptor potential canonical 1, transient receptor potential cation channel, subfamily C, member 1, transient receptor potential channel 1, Transient receptor protein 1, TRP-1, TRP1short transient receptor potential channel 1, TrpC1 | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human TRPC1 (P48995). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title