Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10404R

Description
Thought to form a receptor-activated non-selective calcium permeant cation channel. Probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. Seems to be also activated by intracellular calcium store depletion. Interacts with ITPR3. Appears to be ubiquitous.Specifications
TRPC1 | |
Polyclonal | |
Unconjugated | |
Trpc1 | |
3010009O07Rik; AI115608; capacitative calcium channel protein Trp1; capacitative calcium entry channel; HTRP-1; membe; Mtrp1; mTrp2; Short transient receptor potential channel 1; smTRPC2; transient receptor potential canonical 1; transient receptor potential cation channel subfamily C member 1; transient receptor potential cation channel, subfamily C, member 1; transient receptor potential cation channel, subfamily C, member 1 L homeolog; transient receptor potential protein; Transient receptor protein 1; trp; Trp1; TRP-1; TRP-1 protein; trp2; TRPC1; trpc1.L; TRPC2a; TRPC2b; Trp-related protein 1; Trrp1; Trrp2; XELAEV_18027727mg; xtrpc1 | |
Rabbit | |
Protein A | |
RUO | |
7220 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P48995 | |
Trpc1 | |
KLH conjugated synthetic peptide derived from human TRPC1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction