Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | TRPC3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Description
TRPC3 Polyclonal specifically detects TRPC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
TRPC3 | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence | |
Polyclonal | |
Antisera | |
RUO | |
hTrp3, hTrp-3, short transient receptor potential channel 3, transient receptor potential cation channel, subfamily C, member 3, transient receptor potential channel 3, Transient receptor protein 3, TRP3, TRP-3, TrpC3 | |
TRPC3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Goat | |
Mouse, Human, Rat | |
7222 | |
IgG | |
Unpurified | |
Rat, mouse, human. Other species not yet tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title