Learn More
Invitrogen™ TRPC3 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20258 can be used with blocking peptide PEP-0394.
Specifications
Specifications
| Antigen | TRPC3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Trpc3 |
| Gene Accession No. | Q13507, Q9QZC1 |
| Gene Alias | hTrp3; hTrp-3; ion channel protein; mTrp3; Mwk; receptor-activated cation channel TRP3; SCA41; Short transient receptor potential channel 3; transient receptor potential cation channel subfamily C member 3; transient receptor potential cation channel subfamily C member 3 variant c; transient receptor potential cation channel, subfamily C, member 3; transient receptor protein 3; Trcp3; Trp3; TRP-3; TRPC3; TrpC3c; trp-related protein 3; Trrp3 |
| Gene Symbols | Trpc3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.