Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPC4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5117080

Catalog No. PIPA5117080


Only null left
Add to cart

Description

Description

Positive Samples: SKOV3, BT-474, HeLa, Mouse brain Immunogen sequence: SKESSNSADS DEKSDSEGNS KDKKKNFSLF DLTTLIHPRS AAIASERHNI SNGSALVVQE PPREKQRKVN FVTDIKNFGL FHRRSKQNAA EQNANQIFSV SEEVARQQAA GPLERNIQLE SRGLASRGDL SIPGLSEQCV LVDHRERNTD TLGLQVGKRV CPFKSEKVVV EDTVPIIPKE KHAKEEDSSI DYDLNLPDTV THEDYVTTRL.

TRPC4 is a member of the canonical subfamily of transient receptor potential cation channels. TRPC4 forms a non-selective calcium-permeable cation channel that is activated by Gq-coupled receptors and tyrosine kinases, and plays a role in multiple processes including endothelial permeability, vasodilation, neurotransmitter release and cell proliferation. Single nucleotide polymorphisms in TRPC4 may be associated with generalized epilepsy with photosensitivity. Alternatively spliced transcript variants encoding multiple isoforms have been observed for TRPC4. The classical or canonical transient receptor potential (TRPC) channels are the subfamily most closely related to the founding member of the TRP family, the Drosophila TRP channel. TRPC channels have seven TRPC genes that encode the channel protein (TRPC1-7). Diseases associated with TRPC4 include Photosensitive Epilepsy.CaV3.3 encodes the pore-forming alpha subunit of a voltage gated calcium channel. The encoded protein is a member of a subfamily of calcium channels referred to as is a low voltage-activated, T-type, calcium channel. The channel encoded by CaV3.3 is characterized by a slower activation and inactivation compared to other T-type calcium channels. CaV3.3 may be involved in calcium signaling in neurons. Alternate splicing results in multiple transcript variants of CaV3.3. Voltage-gated calcium channels (CaV) are present in the membrane of most excitable cells and mediate calcium influx in response to depolarization, an proteins such as CaV3.3 regulate intracellular processes such as contraction, secretion, neurotransmission and gene expression. Diseases associated with CACNA1I include Childhood Absence Epilepsy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRPC4
Polyclonal
Unconjugated
Trpc4
alpha TRPC4; beta TRPC4; beta-2 TRPC4; capacitative calcium entry channel 1; capacitative calcium entry channel Trp4; CCE1; HTRP4; HTRP-4; MGC119570; MGC119571; MGC119572; MGC119573; putative capacitative calcium entry channel; receptor-activated cation channel TRP4; Short transient receptor potential channel 4; STRPC4; transient receptor potential cation channel subfamily C member 4; transient receptor potential cation channel, subfamily C, member 4; transient receptor potential channel 4; transient receptor potential protein 4; transient receptor protein 4; Trp4; TRPC 4; Trpc4; Trpc4_v2; trp-related protein 4; Trrp4
Rabbit
Affinity Chromatography
RUO
22066, 7223
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9QUQ5, Q9UBN4
Trpc4
Recombinant fusion protein containing a sequence corresponding to amino acids 768-977 of human TRPC4 (NP_0572631).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.