Learn More
Invitrogen™ TRPC4AP Monoclonal Antibody (3G4)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA521614
Catalog No. PIMA521614
Only null left
Add to Cart
Description
Description
Peptide Sequence: VANEESEHNQ ASIVFPPPGA SEENGLPHTS ARTQLPQSMK IMHEIMYKLE VLYVLCVLLM GRQRNQVHRM IAEFKLIPGL NNLFDKLIWR KHSASALVLH GHNQNCDCSP D.
Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control. The DCX(TRUSS) complex specifically mediates the polyubiquitination and subsequent degradation of MYC. Also participates in the activation of NFKB1 in response to ligation of TNFRSF1A, possibly by linking TNFRSF1A to the IKK signalosome. Involved in JNK activation via its interaction with TRAF2. Also involved in elevation of endoplasmic reticulum Ca(2+) storage reduction in response to CHRM1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRPC4AP
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q8TEL6
TRPC4AP
TRPC4AP (NP_056453, 341 a.a. ~ 451 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA
3G4
Unconjugated
TRPC4AP
4833429F06Rik; C20orf188; D2Ertd113e; hypothetical protein; mFLJ00177; PPP1R158; protein phosphatase 1, regulatory subunit 158; Protein TAP1; Protein TRUSS; Rabex-5/Rin2-interacting protein; short transient receptor potential channel 4 associated protein; short transient receptor potential channel 4-associated protein; TAP1 protein; TNF-receptor; TNF-receptor ubiquitous scaffolding/signaling protein; TP4AP; transient receptor potential cation channel subfamily C member 4 associated protein; transient receptor potential cation channel, subfamily C, member 4 associated protein; transient receptor protein 4, associated protein; TRP4-associated protein; Trp4-associated protein TAP1; Trpc4ap; trpc4-associated protein; Trrp4ap; TRUSS; tumor necrosis factor receptor-associated ubiquitous scaffolding and signaling protein
Mouse
Affinity chromatography
RUO
26133
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.