Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPC4AP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111595
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111595 100 μL
PIPA5111330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111595 Supplier Invitrogen™ Supplier No. PA5111595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIWRKHSASA LVLHGHNQNC DCSPDITLKI QFLRLLQSFS DHHENKYLLL NNQELNELSA ISLKANIPEV EAVLNTDRSL VCDGK Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control. The DCX(TRUSS) complex specifically mediates the polyubiquitination and subsequent degradation of MYC. Also participates in the activation of NFKB1 in response to ligation of TNFRSF1A, possibly by linking TNFRSF1A to the IKK signalosome. Involved in JNK activation via its interaction with TRAF2. Also involved in elevation of endoplasmic reticulum Ca(2+) storage reduction in response to CHRM1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPC4AP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRPC4AP
Gene Accession No. Q8TEL6
Gene Alias 4833429F06Rik; C20orf188; D2Ertd113e; hypothetical protein; mFLJ00177; PPP1R158; protein phosphatase 1, regulatory subunit 158; Protein TAP1; Protein TRUSS; Rabex-5/Rin2-interacting protein; short transient receptor potential channel 4 associated protein; short transient receptor potential channel 4-associated protein; TAP1 protein; TNF-receptor; TNF-receptor ubiquitous scaffolding/signaling protein; TP4AP; transient receptor potential cation channel subfamily C member 4 associated protein; transient receptor potential cation channel, subfamily C, member 4 associated protein; transient receptor protein 4, associated protein; TRP4-associated protein; Trp4-associated protein TAP1; Trpc4ap; trpc4-associated protein; Trrp4ap; TRUSS; tumor necrosis factor receptor-associated ubiquitous scaffolding and signaling protein
Gene Symbols TRPC4AP
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TRPC4AP. Recombinant protein control fragment (Product #RP-107400).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26133
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.