Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPC5 Monoclonal Antibody (N67/15), APC
GREENER_CHOICE

Catalog No. PIMA545477
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545477 Supplier Invitrogen™ Supplier No. MA545477
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45477 was sufficient for detection of TrpC5 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 110kDa.

TRPC5 belongs to the transient receptor family. TRPC5 is one of the seven mammalian TRPC (transient receptor potential channel) proteins. The TRPC5 protein is a multi-pass membrane protein and is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC5 is active alone or as a heteromultimeric assembly with TRPC1, TRPC3, and TRPC4. TRPC5 also interacts with multiple proteins including calmodulin, CABP1, enkurin, Na(+)-H+ exchange regulatory factor (NHERF ), interferon-induced GTP-binding protein (MX1), ring finger protein 24 (RNF24), and SEC14 domain and spectrin repeat-containing protein 1 (SESTD1). TRPC5 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC5 is probably operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases, or G-protein coupled receptors. TRPC5 has also been shown to be calcium-selective, and may also be activated by intracellular calcium store depletion. Diseases associated with TRPC5 include Hypertrophic Pyloric Stenosis and Pyloric Stenosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPC5
Applications Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N67/15
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene TRPC5
Gene Accession No. Q9QX29, Q9UL62
Gene Alias capacitative calcium entry channel 2; CCE2; hTRP5; Htrp-5; mtrp5; PPP1R159; protein phosphatase 1, regulatory subunit 159; short transient receptor potential channel 5; short transient receptor potential channel 5a; transient receptor potential cation channel, subfamily C, member 5a; transient receptor potential cation channel subfamily C member 5; transient receptor potential cation channel subfamily c member 5a; transient receptor potential cation channel, subfamily C, member 5; transient receptor potential cation channel, subfamily C, member 5a; transient receptor potential channel 5; transient receptor protein 5; TRP5; TRP-5; TrpC5; trpc5a; trp-related protein 5; Trrp5; vtransient receptor potential cation channel, subfamily C, member 5; wu:fi73f08
Gene Symbols TRPC5
Host Species Mouse
Immunogen Synthetic peptide amino acids 827-845 of human TrpC5 (also known as short transient receptor potential channel 5, and Htrp5).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140933, 22067, 7224
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.