Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrpC5 Mouse anti-Human, Mouse, Clone: N67/15, MilliporeSigma™
Mouse Monoclonal Antibody
$191.34 - $481.79
Specifications
Antigen | TrpC5 |
---|---|
Clone | N67/15 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN245125
|
MilliporeSigma
MABN245125UG |
25 μg |
Each for $191.34
|
|
|||||
MABN2451100
|
MilliporeSigma
MABN2451100UG |
100 μg |
Each for $481.79
|
|
|||||
Description
TrpC5 Monoclonal antibody specifically detects TrpC5 in Human, Mouse samples. It is validated for Western Blotting, Immunohistochemistry.Specifications
TrpC5 | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG2b κ | |
Protein G purified |
N67/15 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse | |
A linear Peptide corresponding to 19 amino acids from the C-terminal region of human Short transient receptor potential channel 5 (TrpC5). | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title