Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPC5 Polyclonal Antibody
TRPC5 Polyclonal Antibody
Supplier: Thermo Scientific PA595221
Description
TRPC5 Polyclonal Antibody for Western Blot

Specifications
TRPC5 | |
Polyclonal | |
Unconjugated | |
TRPC5 | |
capacitative calcium entry channel 2; CCE2; hTRP5; Htrp-5; mtrp5; PPP1R159; protein phosphatase 1, regulatory subunit 159; short transient receptor potential channel 5; short transient receptor potential channel 5a; transient receptor potential cation channel, subfamily C, member 5a; transient receptor potential cation channel subfamily C member 5; transient receptor potential cation channel subfamily c member 5a; transient receptor potential cation channel, subfamily C, member 5; transient receptor potential cation channel, subfamily C, member 5a; transient receptor potential channel 5; transient receptor protein 5; TRP5; TRP-5; TrpC5; trpc5a; trp-related protein 5; Trrp5; vtransient receptor potential cation channel, subfamily C, member 5; wu:fi73f08 | |
Rabbit | |
Affinity chromatography | |
RUO | |
7224 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q9UL62 | |
TRPC5 | |
E.coli-derived human TRPC5 recombinant protein (Position: K684-L973). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction