Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TRPC6-PAM-C20
GREENER_CHOICESDP

Catalog No. 687550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-755-0 50 mg
68-751-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-755-0 Supplier Tocris Bioscience™ Supplier No. 6875/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective TRPC6 positive allosteric modulator

TRPC6-PAM-C20 is a selective TRPC6 positive allosteric modulator. Selectively activates TRPC6 over other TRP channels. Also activates TRPA1. Induces transient increase in intracellular Ca2+ in HEK cells expressing TRPC6 (EC50 = 2.37 μM). Potentiates OAG (1-oleoyl-1-acetyl-sn-glycerol)-induced Ca2+ influx in human platelets, and enhances OAG-induced platelet aggregation.

Specifications

Chemical Name or Material 3-(3-,4-Dihydro-6,7-dimethoxy-3,3-dimethyl-1-isoquinolinyl)-2H-1-benzopyran-2-one
CAS 667427-75-0
Quantity 50 mg
Molecular Formula C22H21NO4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.