Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPC7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580190
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Mouse Brain Tissue, A549 whole cell, COLO320 whole cell, SKOV whole cell.
TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.
Specifications
TRPC7 | |
Polyclonal | |
Unconjugated | |
TRPC7 | |
capacitative calcium channel TRPC7; hTRP7; likley ortholog of mouse transient receptor potential cation channel, subfamily C, member 7; mTRP7; putative capacitative calcium channel; short transient receptor potential channel 7; transient receptor potential cation channel subfamily C member 7; transient receptor potential cation channel, subfamily C, member 7; transient receptor protein 7; TRP7; TRP-7; TRP7 protein; TRPC7; Trrp8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26946, 57113 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q9HCX4, Q9WVC5 | |
TRPC7 | |
E.coli-derived human TRP 7 recombinant protein (Position: K716-I862). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction