Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142465
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142465 Supplier Invitrogen™ Supplier No. PA5142465
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to cross-react with isoform 1, 2 and 3 (NP_065122.1; NP_001161049.1; NP_001161048.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPC7
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TRPC7
Gene Accession No. Q9WVC5
Gene Alias capacitative calcium channel TRPC7; hTRP7; likley ortholog of mouse transient receptor potential cation channel, subfamily C, member 7; mTRP7; putative capacitative calcium channel; short transient receptor potential channel 7; transient receptor potential cation channel subfamily C member 7; transient receptor potential cation channel, subfamily C, member 7; transient receptor protein 7; TRP7; TRP-7; TRP7 protein; TRPC7; Trrp8
Gene Symbols TRPC7
Host Species Goat
Immunogen Peptide with sequence KLCKSKAKSCEND.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26946, 427641
Target Species Mouse, Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.