Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110795 Supplier Invitrogen™ Supplier No. PA5110795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CEATYLLRQS SINSADGYSL YRYHFNGEEL LFEDTSLSTS PGTGVRKKTC SFRIKEEKDV KTHLVPECQN SLHLSLGTST SATPDGSHLA VDDLKNAEES KLGPDIGISK EDD Highest antigen sequence indentity to the following orthologs: Rat - 65%, Mouse - 61%.

The protein encoded by this gene is similar to the transient receptor potential (Trp) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRPM1
Gene Accession No. Q7Z4N2
Gene Alias 4732499L03Rik; AI606771; CSNB1C; EMBL:CAI30141.1}; long transient receptor potential channel 1; Ltrpc1; melastatin; melastatin 1; melastatin-1; MLSN; MLSN1; transient receptor potential cation channel subfamily M member 1; transient receptor potential cation channel, subfamily M, member 1; transient receptor potential melastatin family; Trpm1; trpm1 {ECO:0000312
Gene Symbols TRPM1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TRPM1. Recombinant protein control fragment (Product #RP-105731).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4308
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.