Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM2 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11081601AF700
Description
TRPM2 Polyclonal specifically detects TRPM2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
TRPM2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
EC 3.6.1.13, EREG1MGC133383, Estrogen-responsive element-associated gene 1 protein, KNP3LTrpC-2, Long transient receptor potential channel 2, LTrpC2, LTRPC2TRPC7transient receptor potential cation channel subfamily M member 2, NUDT9H, NUDT9L1, transient receptor potential cation channel, subfamily M, member 2, Transient receptor potential channel 7, TrpC7 | |
Rabbit | |
181 kDa | |
0.1 mL | |
Apoptosis, Cell Biology, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
7226 | |
Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TRPM2 | |
Synthetic peptide made to a C-terminal portion of the rat TRPM2 protein (within residues 1430-1508). [Swiss-Prot# Q5G856] | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for isoform 1, or the long isoform (TRPM2-L), of TRPM2. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction