Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM2 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.72 - $494.86

Specifications

Antigen TRPM2
Concentration 1.5 mg/mL
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11082364T
SDP
View Documents
Novus Biologicals
NB11082364SS
0.025 mL
Each for $181.72
Only null left
Add to Cart
 
NB11082364
SDP
View Documents
Novus Biologicals
NB11082364
0.1 mL
Each for $494.86
Only null left
Add to Cart
 
Description

Description

TRPM2 Polyclonal specifically detects TRPM2 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TRPM2
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:50
Polyclonal
Rabbit
Apoptosis, Cell Biology, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
PBS and 30% Glycerol with 0.1% Sodium Azide
7226
Synthetic peptide made to an internal portion of the mouse TRPM2 protein (within residues 1200-1300). [Swiss-Prot# Q91YD4]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1.5 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Mouse, Rat
EC 3.6.1.13, EREG1MGC133383, Estrogen-responsive element-associated gene 1 protein, KNP3LTrpC-2, Long transient receptor potential channel 2, LTrpC2, LTRPC2TRPC7transient receptor potential cation channel subfamily M member 2, NUDT9H, NUDT9L1, transient receptor potential cation channel, subfamily M, member 2, Transient receptor potential channel 7, TrpC7
TRPM2
IgG
Affinity Purified
172 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.