Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25830625UL
Description
TRPM2 Polyclonal specifically detects TRPM2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
TRPM2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
EC 3.6.1.13, EREG1MGC133383, Estrogen-responsive element-associated gene 1 protein, KNP3LTrpC-2, Long transient receptor potential channel 2, LTrpC2, LTRPC2TRPC7transient receptor potential cation channel subfamily M member 2, NUDT9H, NUDT9L1, transient receptor potential cation channel, subfamily M, member 2, Transient receptor potential channel 7, TrpC7 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
TRPM2 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IKKMLEVLVVKLPLSEHWALPGGSREPGEMLPRKLKRILRQEHWPSFENLLKCGMEVYKGYMDDPRNTDNAWIETVAVSVHFQDQNDVELNRLNSNLHACDSGASIRWQVVDRRIPLYANHKTLLQK | |
25 μL | |
Apoptosis, Cell Biology, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
7226 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction