Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111058
Description
Immunogen sequence: RTRLEKFISE QTKERGGVAI KIPIVCVVLE GGPGTLHTID NATTNGTPCV VVEGSGRVAD VIAQVANLPV SDITISLIQQ KLSVFFQEMF ETFTESRIVE WTKKIQDIVR RRQLLTVFRE GKDGQQDV.
TRPM2 belongs to the ion transport protein family which includes Sodium, Potassium, Calcium ion channels. This family is characterized by proteins possessing 6 transmembrane helices in which the last two helices flank a loop which determines ion selectivity. TRPM2 is activated by oxidative stress and confers susceptibility to cell death. Further, TRPM2 is a calcium-permeable cation channel that is regulated by free intracellular ADP-ribose. Several alternatively spliced transcript variants of the TRPM2 gene have been described, but their full-length nature is not known.
Specifications
| TRPM2 | |
| Polyclonal | |
| Unconjugated | |
| TRPM2 | |
| 9830168K16Rik; C79133; EC 3.6.1.13; EREG 1; EREG1; Estrogen-responsive element-associated gene 1 protein; KNP 3; KNP3; Long transient receptor potential channel 2; LTRPC 2; LTRPC2; LTrpC-2; MGC133383; NUDT9H; NUDT9L1; null; transient receptor potential cation channel subfamily M member 2; transient receptor potential cation channel, subfamily M, member 2; transient receptor potential channel 7; Transient receptor potential melastatin 2; transient receptor potential melastatin family 2; transient receptor protein 7; Trp7; TRPC 7; TRPC7; TRPM 2; Trpm2; Trpm2-predicted; Trrp7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7226 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.50 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94759 | |
| TRPM2 | |
| Recombinant Protein corresonding to Human TRPM2. Recombinant protein control fragment (Product #RP-103848). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction