Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119946 Supplier Invitrogen™ Supplier No. PA5119946
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: THP-1 cell lysate, PC-3 cell lysate, rat cerebellum tissue, human bone tissue, mouse brain tissue, SH-SY5Y. Subcellular Location: Lysosome, cell membrane, perikaryon, cell projection, cytoplasmic vesicle.

TRPM2 belongs to the ion transport protein family which includes Sodium, Potassium, Calcium ion channels. This family is characterized by proteins possessing 6 transmembrane helices in which the last two helices flank a loop which determines ion selectivity. TRPM2 is activated by oxidative stress and confers susceptibility to cell death. Further, TRPM2 is a calcium-permeable cation channel that is regulated by free intracellular ADP-ribose. Several alternatively spliced transcript variants of the TRPM2 gene have been described, but their full-length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene TRPM2
Gene Accession No. E9PTA2, O94759, Q91YD4
Gene Alias 9830168K16Rik; C79133; EC 3.6.1.13; EREG 1; EREG1; Estrogen-responsive element-associated gene 1 protein; KNP 3; KNP3; Long transient receptor potential channel 2; LTRPC 2; LTRPC2; LTrpC-2; MGC133383; NUDT9H; NUDT9L1; null; transient receptor potential cation channel subfamily M member 2; transient receptor potential cation channel, subfamily M, member 2; transient receptor potential channel 7; Transient receptor potential melastatin 2; transient receptor potential melastatin family 2; transient receptor protein 7; Trp7; TRPC 7; TRPC7; TRPM 2; Trpm2; Trpm2-predicted; Trrp7
Gene Symbols TRPM2
Host Species Rabbit
Immunogen Synthetic peptide within mouse TRPM2 aa 100-150/1503 (Cytoplasmic).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28240, 294329, 7226
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.