Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPM2 Polyclonal Antibody, DyLight™ 488
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522878
Description
TRPM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F)
TRPM2 belongs to the ion transport protein family which includes Sodium, Potassium, Calcium ion channels. This family is characterized by proteins possessing 6 transmembrane helices in which the last two helices flank a loop which determines ion selectivity. TRPM2 is activated by oxidative stress and confers susceptibility to cell death. Further, TRPM2 is a calcium-permeable cation channel that is regulated by free intracellular ADP-ribose. Several alternatively spliced transcript variants of the TRPM2 gene have been described, but their full-length nature is not known.
Specifications
TRPM2 | |
Polyclonal | |
DyLight 488 | |
TRPM2 | |
9830168K16Rik; C79133; EC 3.6.1.13; EREG 1; EREG1; Estrogen-responsive element-associated gene 1 protein; KNP 3; KNP3; Long transient receptor potential channel 2; LTRPC 2; LTRPC2; LTrpC-2; MGC133383; NUDT9H; NUDT9L1; null; transient receptor potential cation channel subfamily M member 2; transient receptor potential cation channel, subfamily M, member 2; transient receptor potential channel 7; Transient receptor potential melastatin 2; transient receptor potential melastatin family 2; transient receptor protein 7; Trp7; TRPC 7; TRPC7; TRPM 2; Trpm2; Trpm2-predicted; Trrp7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28240, 294329 | |
4°C, store in dark | |
Liquid |
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
1.25 mg/mL | |
50 mM sodium borate with no preservative | |
E9PTA2, Q91YD4 | |
TRPM2 | |
Synthetic peptide made to a C-terminal portion of the rat protein (within residues 1430-1508). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction