Learn More
Invitrogen™ TRPM2 Polyclonal Antibody, DyLight™ 550

Description
TRPM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)
Specifications
Specifications
| Antigen | TRPM2 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.83 mg/mL |
| Conjugate | DyLight 550 |
| Formulation | 50mM sodium borate with 0.05% sodium azide |
| Gene | TRPM2 |
| Gene Accession No. | E9PTA2, Q91YD4 |
| Gene Alias | 9830168K16Rik; C79133; EC 3.6.1.13; EREG 1; EREG1; Estrogen-responsive element-associated gene 1 protein; KNP 3; KNP3; Long transient receptor potential channel 2; LTRPC 2; LTRPC2; LTrpC-2; MGC133383; NUDT9H; NUDT9L1; null; transient receptor potential cation channel subfamily M member 2; transient receptor potential cation channel, subfamily M, member 2; transient receptor potential channel 7; Transient receptor potential melastatin 2; transient receptor potential melastatin family 2; transient receptor protein 7; Trp7; TRPC 7; TRPC7; TRPM 2; Trpm2; Trpm2-predicted; Trrp7 |
| Gene Symbols | TRPM2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.