Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$207.00 - $499.50
Specifications
Antigen | TRPM3 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017958
![]() |
Novus Biologicals
NB100988640.025ML |
0.025 mL |
Each for $207.00
|
|
|||||
NB10098864
![]() |
Novus Biologicals
NB10098864 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
TRPM3 Polyclonal specifically detects TRPM3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TRPM3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
EC 2.3.1.31, EC 5.99.1.3, Long transient receptor potential channel 3, LTrpC3, LTrpC-3, LTRPC3KIAA1616MLSN2GON-2, melastatin 2, melastatin-2, transient receptor potential cation channel subfamily M member 3, transient receptor potential cation channel, subfamily M, member 3 | |
TRPM3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
Q9HCF6 | |
80036 | |
IgG | |
Unpurified | |
Specific for TRPM3. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title