Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139988 Supplier Invitrogen™ Supplier No. PA5139988
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VDIRLAQLED LIGRMATALE RLTGLERAES NKIRSRTSSD CTDAAYIVRQ SSFNSQEGNT FKLQ Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The product of this gene belongs to the family of transient receptor potential (TRP) channels. TRP channels are cation-selective channels important for cellular calcium signaling and homeostasis. The protein encoded by this gene mediates calcium entry, and this entry is potentiated by calcium store depletion. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRPM3
Gene Accession No. Q9HCF6
Gene Alias 6330504P12Rik; 9330180E14; AU018608; B930001P07Rik; GON-2; KI; KIAA1616; Long transient receptor potential channel 3; LTRPC3; LTrpC-3; melastatin 2; melastatin-2; MLSN2; si:dkey-201c13.4; transient receptor potential cation channel subfamily M member 3; transient receptor potential cation channel, subfamily M, member 3; transient receptor potential melastatin 3; transient receptor potential melastatin 3 beta 1; transient receptor potential melastatin 3 beta 10; transient receptor potential melastatin 3 beta 11; transient receptor potential melastatin 3 beta 12; transient receptor potential melastatin 3 beta 13; transient receptor potential melastatin 3 beta 14; transient receptor potential melastatin 3 beta 15; transient receptor potential melastatin 3 beta 16; transient receptor potential melastatin 3 beta 17; transient receptor potential melastatin 3 beta 2; transient receptor potential melastatin 3 beta 3; transient receptor potential melastatin 3 beta 4; transient receptor potential melastatin 3 beta 5; transient receptor potential melastatin 3 beta 6; transient receptor potential melastatin 3 beta 7; transient receptor potential melastatin 3 beta 8; transient receptor potential melastatin 3 beta 9; trpm3
Gene Symbols TRPM3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRPM3. Recombinant protein control fragment (Product #RP-109408).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.