Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRPM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116483
Description
Positive controls: SK-Br-3 cell lysates, rat heart tissue, mouse small intestine tissue, MCF-7.
Calcium-activated non selective (CAN) cation channel that mediates membrane depolarization. While it is activated by increase in intracellular Ca2+, it is impermeable to it. Mediates transport of monovalent cations (Na+, K+, Cs+, Li+), leading to depolarize the membrane. It thereby plays a central role in cadiomyocytes, neurons from entorhinal cortex, dorsal root and vomeronasal neurons, endocrine pancreas cells, kidney epithelial cells, cochlea hair cells etc. Participates in T-cell activation by modulating Ca2+ oscillations after T lymphocyte activation, which is required for NFAT-dependent IL2 production. Involved in myogenic constriction of cerebral arteries. Controls insulin secretion in pancreatic beta-cells. May also be involved in pacemaking or could cause irregular electrical activity under conditions of Ca2+ overload. Affects T-helper 1 (Th1) and T-helper 2 (Th2) cell motility and cytokine production through differential regulation of calcium signaling and NFATC1 localization. Enhances cell proliferation through up-regulation of the beta-catenin signaling pathway.
Specifications
| TRPM4 | |
| Polyclonal | |
| Unconjugated | |
| TRPM4 | |
| 1110030C19Rik; AW047689; Calcium-activated non-selective cation channel 1; H LTrpC-4; hTRPM4; Long transient receptor potential channel 4; LOW QUALITY PROTEIN: transient receptor potential cation channel subfamily M member 4; LTrpC4; LTrpC-4; melastatin like 2 protein; melastatin-4; Melastatin-like 2; MLS2s; PFHB1B; transient receptor potential cation channel subfamily M member 4; transient receptor potential cation channel, subfamily M, member 4; transient receptor potential ion channel melastatin subgroup member 4; Trpm4; TRPM4B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171143, 54795, 68667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q7TN37, Q8TD43, Q9ESQ5 | |
| TRPM4 | |
| Synthetic peptide within human TRPM4 aa 1-50. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction