Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580191
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580191 Supplier Invitrogen™ Supplier No. PA580191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: SMMC whole cell, 293T whole cell. IHC: mouse brain tissue, rat testis tissue, human prostatic cancer tissue.

This gene encodes a member of the transient receptor potential protein family, which is a diverse group of proteins with structural features typical of ion channels. This protein plays an important role in taste transduction, and has characteristics of a calcium-activated, non-selective cation channel that carries Na+, K+, and Cs+ ions equally well, but not Caions. It is activated by lower concentrations of intracellular Ca, and inhibited by higher concentrations. It is also a highly temperature-sensitive, heat activated channel showing a steep increase of inward currents at temperatures between 15 and 35 degrees Celsius. This gene is located within the Beckwith-Wiedemann syndrome critical region-1 on chromosome 11p15. 5, and has been shown to be imprinted, with exclusive expression from the paternal allele.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene trpm5
Gene Accession No. Q9JJH7, Q9NZQ8
Gene Alias 9430099A16Rik; Long transient receptor potential channel 5; long transient receptor potential-related channel 5; LTRPC5; LTrpC-5; MLSN1 and TRP-related; MLSN1- and TRP-related gene 1 protein; MTR1; si:dkey-56p7.1; transient receptor potential cation channel subfamily M member 5; transient receptor potential cation channel, subfamily M, member 5; transient receptor potential melastatin 5; TRPM5
Gene Symbols trpm5
Host Species Rabbit
Immunogen E.coli-derived human TRPM5 recombinant protein (Position: S1021-Q1108).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29850, 365391, 56843
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.