Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988781
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8781 Supplier Bioss Supplier No. BS9048R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPM6 is predominantly expressed in the kidney and colon, and encodes a protein containing an ion channel domain and a protein kinase domain. TRPM6 is crucial for magnesium homeostasis, and plays an essential role in epithelial magnesium transport and in the active magnesium absorption in the gut and kidney. Further, TRPM6 is an essential ion channel and serine/threonine-protein kinase, and is crucial for magnesium homeostasis. TRPM6 has an important role in epithelial magnesium transport and in the active magnesium absorption in the gut and kidney. Isoforms of the type M6-kinase lack the ion channel region. Mutations in TRPM6 are associated with hypomagnesemia with secondary hypocalcemia. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene. TRPC7 is thought to form a receptor-activated non-selective calcium permeant cation channel. TRPC7 is probably is operated by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases or G-protein coupled receptors. TRPC7 is also activated by diacylglycerol (DAG). Further, TRPC7 may also be activated by intracellular calcium store depletion.

Specifications

Antigen TRPM6
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Trpm6
Gene Accession No. Q9BX84
Gene Alias CHAK2; Channel kinase 2; HMGX; HOMG; HOMG1; HSH; Melastatin-related TRP cation channel 6; transient receptor potential cation channel 6/channel-kinase 2; Transient receptor potential cation channel subfamily M member 6; transient receptor potential cation channel, subfamily M, member 6; Trpm6; TRPM6/CHAK2
Gene Symbols Trpm6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TRPM6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140803
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.