Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM7 Antibody (SR1322), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32185250UL
Description
TRPM7 Monoclonal antibody specifically detects TRPM7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ ImmunofluorescenceSpecifications
TRPM7 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1322 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK | |
A synthesized peptide derived from human TRPM7 (Uniprot #: Q96QT4 ) | |
50 μg | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
54822 | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction