Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM7, p Ser1513 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323186
Description
TRPM7 Polyclonal antibody specifically detects TRPM7 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TRPM7 | |
Polyclonal | |
Western Blot 1:500 | |
CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK | |
Phospho-TRPM7 (Ser-1513) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding Ser-1513 in human TRPM7. This site is well conserved in rat and mouse TRPM7, but has low homology to other TRPM family members. [UniProt# Q96QT4] | |
0.1 mL | |
Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
54822 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction