Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPM7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102835 Supplier Invitrogen™ Supplier No. PA5102835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TRPM7.

TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Trpm7
Gene Accession No. Q923J1, Q925B3, Q96QT4
Gene Alias 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4}
Gene Symbols Trpm7
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRPM7(Accession Q96QT4), corresponding to amino acid residues D1832-L1865.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54822, 58800, 679906
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.