Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM8 Antibody (M572), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP323238
Description
TRPM8 Monoclonal antibody specifically detects TRPM8 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
TRPM8 | |
Monoclonal | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Mouse | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
M572 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200 | |
Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
Clone M572 was generated from TRPM8 synthetic peptide (coupled to carrier) corresponding to amino acids in the extracellular region of human TRPM8. This site is well conserved in rat and mouse TRPM8, but has low homology to other TRPM family members. [UniProt# Q7Z2W7] | |
0.1 mL | |
Cancer, Tumor Suppressors | |
79054 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction