Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRPM8 Antibody (SR1110), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321832100UL
Description
TRPM8 Monoclonal antibody specifically detects TRPM8 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
TRPM8 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunofluorescence | |
SR1110 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8 | |
A synthesized peptide derived from human TRPM8 (Uniprot #: Q7Z2W7) | |
100 μg | |
Cancer, Tumor Suppressors | |
79054 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction