Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPM8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA14282

Catalog No. PA14282


Only null left
Add to Cart

Description

Description

PA1-4282 detects the Trp-p8 protein in human samples. PA1-4282 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~125 kDa protein representing Trp-p8 in transfected COS cells. PA1-4282 immunizing peptide corresponds to amino acid residues 278-292 and the C terminus sequence of the human Trp-p8 protein.

TRPM8 is a receptor-activated non-selective cation channel involved in detection of sensations such as coolness, by being activated by cold temperature below 25 degrees Celsius. TRPM8 is activated by icilin, eucalyptol, menthol, cold and modulation of intracellular pH and involved in menthol sensation. TRPM8 is permeable for monovalent cations sodium, potassium, and cesium and divalent cation calcium. TRPM8 has an expression pattern within normal human tissues that is mostly restricted to prostate epithelial cells. TRPM8 mRNA expression is at moderate levels in normal prostate tissue and appears to be elevated in prostate cancer. Notably, TRPM8 mRNA was also expressed in a number of nonprostatic primary tumors of breast, colon, lung, and skin origin, whereas transcripts encoding TRPM8 were hardly detected or not detected in the corresponding normal human tissues.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRPM8
Polyclonal
Unconjugated
TRPM8
CMR1; Cold menthol receptor 1; cold/menthol receptor 1; long transient receptor potential channel 6; LTRPC6; LTrpC-6; MGC2849; short form of the TRPM8 cationic channel; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; transient receptor potential p8; transient receptor potential subfamily M member 8; transient receptor potential-p8; TRP ion channel TRPM8; Trpm8; Trpp8; trp-p8
Rabbit
Antigen Affinity Chromatography
RUO
79054
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q7Z2W7
TRPM8
Synthetic peptide: R(278) N Q L E K Y I S E R T I Q D(292) and synthetic peptide to the C terminus: (1090) N D L K G L L K E I A N K I K(1104).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.