Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPM8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534287
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534287 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534287 Supplier Invitrogen™ Supplier No. PA534287
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Baboon, Monkey, Marmoset, Mouse, Rat, Guinea pig (100%).

TRPM8 is a receptor-activated non-selective cation channel involved in detection of sensations such as coolness, by being activated by cold temperature below 25 degrees Celsius. TRPM8 is activated by icilin, eucalyptol, menthol, cold and modulation of intracellular pH and involved in menthol sensation. TRPM8 is permeable for monovalent cations sodium, potassium, and cesium and divalent cation calcium. TRPM8 has an expression pattern within normal human tissues that is mostly restricted to prostate epithelial cells. TRPM8 mRNA expression is at moderate levels in normal prostate tissue and appears to be elevated in prostate cancer. Notably, TRPM8 mRNA was also expressed in a number of nonprostatic primary tumors of breast, colon, lung, and skin origin, whereas transcripts encoding TRPM8 were hardly detected or not detected in the corresponding normal human tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPM8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TRPM8
Gene Accession No. Q7Z2W7, Q8R455, Q8R4D5
Gene Alias CMR1; Cold menthol receptor 1; cold/menthol receptor 1; long transient receptor potential channel 6; LTRPC6; LTrpC-6; MGC2849; short form of the TRPM8 cationic channel; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; transient receptor potential p8; transient receptor potential subfamily M member 8; transient receptor potential-p8; TRP ion channel TRPM8; Trpm8; Trpp8; trp-p8
Gene Symbols TRPM8
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from internal region of human TRPM8.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100337634, 171382, 171384, 79054
Target Species Human, Mouse, Rat, Monkey, Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.