Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen TRPM8
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125629
SDP
View Documents
Novus Biologicals
NBP310364100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

TRPM8 Polyclonal specifically detects TRPM8 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

TRPM8
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Cancer, Tumor Suppressors
PBS buffer, 2% sucrose
79054
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml, Immunohistochemistry
Polyclonal
Purified
RUO
Human
Long transient receptor potential channel 6, LTrpC-6, LTRPC6, MGC2849, short form of the TRPM8 cationic channel, transient receptor potential cation channel, subfamily M, member 8, Transient receptor potential p8, transient receptor potential subfamily M member 8, trp-p8, TRPP8transient receptor potential cation channel subfamily M member 8
The immunogen is a synthetic peptide directed towards the N terminal region of human TRPM8 (NP_076985). Peptide sequence YILDNNHTHLLLVDNGCHGHPTVEAKLRNQLEKYISERTIQDSNYGGKIP
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.