Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPML1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111029
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111029 Supplier Invitrogen™ Supplier No. PA5111029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTIKHPGGAG AEESELQAYI AQCQDSPTSG KFRRGSGSAC SLLCCCGRDP SE Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 85%.

Defects in Mucolipin-1 are the cause of mucolipidosis type IV (MLIV), also known as sialolipidosis. MLIV is an autosomal recessive lysosomal storage disorder characterized by severe psychomotor retardation and ophthalmologic abnormalities, including corneal opacity, retinal degeneration and strabismus. Storage bodies of lipids and water-soluble substances are seen by electron microscopy in almost every cell type of the patients. Most patients are unable to speak or walk independently and reach a maximal developmental level of 1-2 years. All patients have constitutive achlorhydia associated with a secondary elevation of serum gastrin levels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPML1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCOLN1
Gene Accession No. Q9GZU1
Gene Alias 2210015I05Rik; MCLN1; Mcoln1; MCOLN1 mucolipin 1; MG-2; MGC7172; ML1; ML4; MLIV; MST080; MSTP080; Mucolipidin; mucolipidosis type IV protein; mucolipin 1; mucolipin-1; Transient receptor potential channel mucolipin 1; Transient receptor potential-mucolipin 1; TRPML1; TRP-ML1; TRPM-L1
Gene Symbols MCOLN1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TRPML1. Recombinant protein control fragment (Product #RP-95585).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57192
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.