Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPML2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116860 Supplier Invitrogen™ Supplier No. PA5116860
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: THP-1, SKOV3 Immunogen sequence: VVAFKEDNTV AFKHLFLKGY SGTDEDDYSC SVYTQEDAYE SIFFAINQYH QLKDITLGTL GYGENEDNRI GLKVCKQHYK KGTMFPSNET LNIDNDVELD CVQLDLQDLS KKPPDWKNSS FFRLEFYRLL QVEISFHLKG IDLQTIHSRE LPDCYVFQNT IIFDNKAHSG KIKIYFDSDA KIEECK.

Mucolipins constitute a family of cation channel proteins with homologs in mouse, Drosophila, and C. elegans. Mutations in the human MCOLN1 gene cause mucolipodosis IV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPML2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.90 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MCOLN2
Gene Accession No. Q8IZK6
Gene Alias 3300002C04Rik; AI549968; C86638; FLJ36691; MCLN2; MCOLN2; mucolipidin 2; mucolipin 2; mucolipin-2; Transient receptor potential channel mucolipin 2; TRPML2; TRP-ML2
Gene Symbols MCOLN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-275 of human MCOLN2 (NP_6949912).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255231, 292168
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.