Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPML3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553725
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553725 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553725 Supplier Invitrogen™ Supplier No. PA553725
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YENKGTKQSA MAICQHFYKR GNIYPGNDTF DIDPEIETEC FFVEPDEPFH IGTPAENKLN LTLDFHRLLT VELQFKLKAI NLQTVRHQEL PDCYDFTLT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.

Expressed in the cochlea; particularly in the inner and outer hair cells. Defects in Mcoln3 are the cause of the varitin-waddler (Va) phenotype. Classical Va mice exhibit early-onset hearing loss, vestibular defects, pigmentation abnormalities and perinatal lethality. The phenotype varitin-waddler Jackcon (Va-J), which arose in a cross segregating for Va, is similar but less severe. Belongs to the transient receptor family, polycystin subfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPML3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MCOLN3
Gene Accession No. Q8TDD5
Gene Alias 6720490O21Rik; FLJ11006; FLJ36629; MCLN3; Mcoln3; Mcoln3 mucolipin 3; MGC124245; MGC124246; MGC71509; mucolipin 3; mucolipin-3; Transient receptor potential channel mucolipin 3; TRPML3; TRP-ML3; Va; varitint-waddler
Gene Symbols MCOLN3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Mucolipin 3. Recombinant protein control fragment (Product #RP-91207).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55283
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.